Learn More
Texwipe™ VersaWipe™ Wipers
For ISO Class 6 to 7 (Class 1000 to 10,000) applications
$35.55 - $443.68
Specifications
Material Polyester Cellulose
Autoclavable Autoclavable
Basis Weight 60
View More Specs
Products 3
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity  
19-150-0105
View Documents
Texwipe™
TX624
4 x 4 in. (10.16 x 10.16 cm)
Case of 10 Each for $443.68
Only null left
19-120-2433
View Documents
Texwipe™
TX629
9 x 9 in. (22.86 x 22.86 cm)
Pack of 300 Each for $35.55
Case of 10 Pack for $355.50
Only null left
19-130-3863
View Documents
Texwipe™
TX622
12 x 12 in.
Case of 1500 Each for $342.85
Only null left
Description
Description
Highly absorbent, durable wipers made of a nonwoven blend of 55% cellulose and 45% polyester.
  • Blend of 55% cellulose and 45% polyester offers low extractable levels and particle count
  • Economical wiper is ideal general wiping and spill control in controlled environments
  • Compatible with solvents
  • Contain no chemical binders
  • Excellent wet and dry strength
  • Autoclavable
Order Info
9 x 9 in. wipers available in packs of 300 and case of 10/pack. 12 x 12 in. wipers available 10 bags/case (150 wipers/bag).
Specifications
Specifications
Polyester Cellulose
Autoclavable
60