Learn More
Streptavidin HRP Conjugate , TCI America™
Purified streptavidin, Horseradish Peroxidase (HRP) conjugated.
Supplier:  TCI America S09721VIAL
Catalog No. S0972VIAL
Only null left
Explore available promotions
Description
Description
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications
Specifications
HRP
0.1 mg
Store at -20°C to -80°C (preferably at -80°C). It is not recommended storing at 4°C. Avoid repeated freezing and thawing.
Streptavidin
Product Suggestions
Product Suggestions