missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ XRF Spec-Caps™ Cups

These XRF Spec-Caps™ Cups aid in the production of pressed powder sample disks for XRF, OES, and other analytical techniques.

Supplier:  Cole-Parmer™ 3617M

 View more versions of this product

Catalog No. 04-675-20


Only null left
Explore available promotions

Description

Description

Aluminum cups aid in the production of pressed-powder sample discs for XRF.

  • Reinforces sample disks to resist chipping and breaking.
  • Makes sample disks easier to remove from the pellet die.
  • Minimizes contamination during sample handling.
  • Protects the bore of the pellet die from abrasive samples.
  • Shallow, thin-walled aluminum cups form the bottom and sides of the finished pellet to help reinforce XRF pellets.
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications

Specifications

XRF Spectrometers
Produces pellets 40 mm x 5 mm
38 mm
0.35 in.
1000 Pk.
Sample Cup
1.5 in.
Sample preparation
9 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions