missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ SAF-T-FIT™ Plus N95 Particulate Respirators
GREENER_CHOICE

For use with particulate aerosols free of oil

$26.96 - $557.24

Specifications

Aerosol Type N - Not Resistant to Oil
Design Type Cup/Molded
Disposable Single-use
Filter Type N95
Filter Efficiency 95%
View More Specs

Products 8
Catalog Number Mfr. No. With Exhalation Valve Color Product Type Size Price Quantity  
Catalog Number Mfr. No. With Exhalation Valve Color Product Type Size Price Quantity  
29-635-5348
View Documents
Honeywell™
14110395
Yes White General Purpose Particulate Respirator X-Large
Case of 200 Each for $557.24
Only null left
 
29-635-5345
View Documents
Honeywell™
14110392
No White General Purpose Particulate Respirator X-Large
Pack of 20 Each for $30.39
Case of 10 Pack for $289.90
Only null left
 
29-635-5343
View Documents
Honeywell™
14110390
No White General Purpose Particulate Respirator Small
Pack of 20 Each for $30.88
Case of 10 Pack for $294.50
Only null left
 
29-635-5344
View Documents
Honeywell™
14110391
No White General Purpose Particulate Respirator Medium/Large
Pack of 20 Each for $27.07
Case of 10 Pack for $81.50
Only null left
 
29-635-5340
View Documents
Honeywell™
14110387
No White General Purpose Particulate Respirator Small
Pack of 20 Each for $26.96
Case of 10 Pack for $70.40
Only null left
 
29-635-5349
View Documents
Honeywell™
14110396
Yes Gray Particulate Respirator with Nuisance Level Organic Vapor Relief Small
Pack of 10 Each for $61.39
Case of 10 Pack for $585.60
Only null left
 
29-635-5351
View Documents
Honeywell™
14110398
Yes Gray Particulate Respirator with Nuisance Level Organic Vapor Relief X-Large
Pack of 10 Each for $59.42
Case of 10 Pack for $100.40
Only null left
 
29-635-5346
View Documents
Honeywell™
14110393
Yes White General Purpose Particulate Respirator Small
Pack of 20 Each for $63.66
Case of 10 Pack for $607.20
Only null left
 
Description

Description

  • Low breathing resistance
  • Incorporates humidity-resistant filter media
  • Durable suspension straps
  • Packaged to maintain integrity of product in work environment
  • Adjustable boomerang nose seal for secure fit
  • Color coded nose clips
  • Latex-free

Specifications

Specifications

N - Not Resistant to Oil
Single-use
95%
Two-Strap Design
Cup/Molded
N95
Yes
No
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'