missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ SAF-T-FIT™ Plus N95 Particulate Respirators

For use with particulate aerosols free of oil

Supplier:  Honeywell™ 14110390

 View more versions of this product

Catalog No. 29-635-5343



Only null left
Explore available promotions

Description

Description

  • Low breathing resistance
  • Incorporates humidity-resistant filter media
  • Durable suspension straps
  • Packaged to maintain integrity of product in work environment
  • Adjustable boomerang nose seal for secure fit
  • Color coded nose clips
  • Latex-free

Specifications

Specifications

N - Not Resistant to Oil
White
Single-use
General Purpose Particulate Respirator
No
Welding, soldering, brazing, torch cutting, metal pouring, grinding, sanding
14110390
Two-Strap Design
NIOSH 42 CFR 84, N95, NIOSH approval number: TC-84A-3769
Cup/Molded
N95
Small
95%
Yes
20/Pk.
No
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions