missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ Sperian™ SAF-T-FIT™ P95 Particulate Respirators
For use with contaminant aerosols containing oil
Supplier: Honeywell™ 14110430
Description
- Low breathing resistance
- Incorporate humidity-resistant filter media
- Durable suspension straps
- Packaged to maintain integrity of product in work environment
- Adjustable boomerang nose seal for secure fit
- Color-coded nose clips
- Latex-free
- P1135 and P1135-1 feature full-sealing flange and adjustable straps
Specifications
P - Oil-Proof | |
Cup/Molded | |
P95 | |
Medium/Large | |
95% | |
14110430 | |
Suspension Straps |
NIOSH 42 CFR 84, P95, approval number: TC-84A-3879 | |
Single-use | |
Particulate Respirator with Nuisance Organic Vapor Relief | |
Yes | |
Yes | |
10/Pk. | |
No |