missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Honeywell™ Sperian™ SAF-T-FIT™ P95 Particulate Respirators
For use with contaminant aerosols containing oil
$73.77 - $73.77
Specifications
Certifications/Compliance | NIOSH TC-84A-3879 |
---|---|
Product Type | P1135-1 |
Quantity | 10/Pk. |
Catalog Number | Mfr. No. | Certifications/Compliance | Product Type | Quantity | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Certifications/Compliance | Product Type | Quantity | Price | Quantity | |||
29-635-5362
|
Honeywell™
14110430 |
NIOSH TC-84A-3879 | P1135-1 | 10/Pk. |
Case of 10 Each for $73.77
|
|
Description
- Low breathing resistance
- Incorporate humidity-resistant filter media
- Durable suspension straps
- Packaged to maintain integrity of product in work environment
- Adjustable boomerang nose seal for secure fit
- Color-coded nose clips
- Latex-free
- P1135 and P1135-1 feature full-sealing flange and adjustable straps
Specifications
NIOSH TC-84A-3879 | |
P1135-1 | |
10/Pk. |