missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Wide Mouth Color-Code Safety Labeled Wash Bottles

Easy-to-read labeling printed bottle promoting safe working environment

Supplier:  Bel-Art™ F116460611

 View more versions of this product

Catalog No. 22-273739


Only null left
Explore available promotions

Includes: Labeling with Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols, Chemical Abstract Service (CAS) number, protective equipment symbols and target organ information

Description

Description

Easy-to-read labeling printed bottle promoting safe working environment

  • Wide mouth 53 mm (2 in.) bottle for easy refilling.
  • Made from solvent-resistant low-density polyethylene.
  • Polypropylene closure with leakproof design.
  • Pictogram instructions promote universal safety regardless of setting.
  • Use LYOB only with solvents compatible with LDPE.

Compliance

Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200

Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications

Specifications

16 oz., 500 mL
16 oz.
Methyl Ethyl Ketone
Non-sterile
Yes
Reusable
Wide
Wash Bottle
Polypropylene
Polypropylene Closure
500 mL
Clear
LDPE
OSHA 29 CFR 1910.1200
Not Autoclavable
No
Cylindrical
Methyl Ethyl Ketone
53 mm
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions