missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ Wide Mouth Color-Code Safety Labeled Wash Bottles

Easy-to-read labeling printed bottle promoting safe working environment
$135.47 - $391.43
Specifications
| Color | Clear |
|---|---|
| Sterility | Non-sterile |
| Closure Included | Yes |
| Disposable | Reusable |
| Graduated | No |
| Catalog Number | Mfr. No. | Capacity (Metric) | For Use With (Application) | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | For Use With (Application) | Price | Quantity & Availability | ||||
22273735
![]() |
Bel-Art™
F116466250 |
500 mL | Dichloromethane |
Case of 12 Each for $391.43
|
|
||||
22288641
![]() |
Bel-Art™
F116460640 |
500 mL | Ethanol |
Case of 12 Each for $252.50
|
|
||||
22273739
![]() |
Bel-Art™
F116460611 |
500 mL | Methyl Ethyl Ketone |
Pack of 6 Each for $135.47
|
|
||||
1451238
![]() |
Bel-Art™
F116460627 |
500 mL | Sodium Hypochlorite |
Pack of 6 Each for $144.27
Case of 2 Pack for $288.54
|
|
||||
22288648
![]() |
Bel-Art™
F116461132 |
1000 mL | Methyl Ethyl Ketone |
Pack of 6 Each for $376.30
|
|
||||
22288650
![]() |
Bel-Art™
F116462232 |
1000 mL | Acetone |
Pack of 6 Each for $202.79
Case of 2 Pack for $405.58
|
|
||||
Description
Easy-to-read labeling printed bottle promoting safe working environment
- Wide mouth 53 mm (2 in.) bottle for easy refilling.
- Made from solvent-resistant low-density polyethylene.
- Polypropylene closure with leakproof design.
- Pictogram instructions promote universal safety regardless of setting.
- Use LYOB only with solvents compatible with LDPE.
Compliance
Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
Specifications
| Clear | |
| Yes | |
| No | |
| Cylindrical | |
| Not Autoclavable |
| Non-sterile | |
| Reusable | |
| Wide | |
| Polypropylene |