missing translation for 'onlineSavingsMsg'
Learn More

Sodium Polyphosphates, Glassy, FCC, 60-71%, Spectrum™ Chemical

SO169, 68915-31-1, Approx. (NaPO3)n

Supplier:  Spectrum Chemical Mfg Cor SO16912KG

 View more versions of this product

Catalog No. 18606746


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Sodium Polyphosphates, Glassy, FCC is used as an acidity regulator, emulsifier and buffering agent, and as a sequestering agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Specifications

68915-31-1
12 kg
GYQBBRRVRKFJRG-UHFFFAOYSA-L
disodium hydrogen (hydrogen phosphonatooxy)phosphonate
60 to 71%
Poly Pail
1
H2Na2O7P2
[Na+].[Na+].OP([O-])(=O)OP(O)([O-])=O
221.94
FCC

Chemical Identifiers

68915-31-1
221.94
disodium hydrogen (hydrogen phosphonatooxy)phosphonate
H2Na2O7P2
GYQBBRRVRKFJRG-UHFFFAOYSA-L
[Na+].[Na+].OP([O-])(=O)OP(O)([O-])=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions