missing translation for 'onlineSavingsMsg'
Learn More

Sodium Polyphosphates, Glassy, FCC, 60-71%, Spectrum™ Chemical

Catalog No. p-7158261
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Catalog No. Quantity Packaging
18606745 2.5 kg Poly Bottle
18606746 12 kg Poly Pail
18606747 45 kg Fiber Drum
3 options

Catalog No. 18606745

Supplier: Spectrum Chemical Mfg Cor SO1692.5KG

Only null left

SO169, 68915-31-1, Approx. (NaPO3)n

Spectrum™ Chemical Sodium Polyphosphates, Glassy, FCC is used as an acidity regulator, emulsifier and buffering agent, and as a sequestering agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 68915-31-1
Molecular Formula H2Na2O7P2
Molecular Weight (g/mol) 221.94
InChI Key GYQBBRRVRKFJRG-UHFFFAOYSA-L
IUPAC Name disodium hydrogen (hydrogen phosphonatooxy)phosphonate
SMILES [Na+].[Na+].OP([O-])(=O)OP(O)([O-])=O

Specifications

CAS Max % 1
Quantity 2.5 kg
Percent Purity 60 to 71%
Grade FCC
Packaging Poly Bottle