missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Lactate Solution, FCC, 60%, Spectrumâ„¢ Chemical
S1217, 72-17-3, C3H5NaO3
$510.89 - $7,145.57
Chemical Identifiers
Molecular Formula | C3H5NaO3 |
---|---|
Molecular Weight (g/mol) | 112.06 |
InChI Key | NGSFWBMYFKHRBD-UHFFFAOYNA-M |
IUPAC Name | sodium 2-hydroxypropanoate |
SMILES | [Na+].CC(O)C([O-])=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
18606605
|
Spectrum Chemical Mfg Cor
S1217500ML |
500 mL | Amber Glass Bottle |
N/A
|
|
|||||
18606606
|
Spectrum Chemical Mfg Cor
S12174LT |
4 L | Amber Glass Bottle |
N/A
|
|
|||||
18606607
|
Spectrum Chemical Mfg Cor
S121720LT |
20 L | Poly Drum |
N/A
|
|
|||||
18606608
|
Spectrum Chemical Mfg Cor
S1217200LTBL |
200 L | Drum |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | ||||||||||
Description
Spectrumâ„¢ Chemical Sodium Lactate Solution, 60 Percent, FCC is used in food products as a preservative. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
C3H5NaO3 | |
NGSFWBMYFKHRBD-UHFFFAOYNA-M | |
[Na+].CC(O)C([O-])=O |
112.06 | |
sodium 2-hydroxypropanoate |
Specifications
5.0 to 9.0 | |
C3H5NaO3 | |
NGSFWBMYFKHRBD-UHFFFAOYNA-M | |
sodium 2-hydroxypropanoate | |
58.8 to 61.2% |
Amber Glass Bottle | |
500 mL | |
[Na+].CC(O)C([O-])=O | |
112.06 | |
FCC |