missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Lactate Solution, FCC, 60%, Spectrum™ Chemical
S1217, 72-17-3, C3H5NaO3
Supplier: Spectrum Chemical Mfg Cor S121720LT
Description
Spectrum™ Chemical Sodium Lactate Solution, 60 Percent, FCC is used in food products as a preservative. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 5.0 to 9.0 | |
| C3H5NaO3 | |
| NGSFWBMYFKHRBD-UHFFFAOYNA-M | |
| sodium 2-hydroxypropanoate | |
| 58.8 to 61.2% |
| Poly Drum | |
| 20 L | |
| [Na+].CC(O)C([O-])=O | |
| 112.06 | |
| FCC |
Chemical Identifiers
| C3H5NaO3 | |
| NGSFWBMYFKHRBD-UHFFFAOYNA-M | |
| [Na+].CC(O)C([O-])=O |
| 112.06 | |
| sodium 2-hydroxypropanoate |