Learn More
Sodium Hydroxide, Flakes, Technical, Spectrum™ Chemical
S1308, 1310-73-2, NaOH
Supplier:  Spectrum Chemical Mfg Cor S130825KG
 View more versions of this product
Catalog No. 18606561
Only null left
Explore available promotions
Description
Description
Spectrum™ Chemical Sodium Hydroxide, Flakes, Technical is a highly caustic metallic base combined with alkali salt. Commonly known as Caustic soda, this inorganic compound is water soluble, as well as in ethanol and methanol. The dissolution of solid Sodium Hydroxide in water creates a highly exothermic reaction (creates a lot of heat). One of its many characteristics is that it's a deliquescent alkali and absorbs carbon dioxide and moisture from the air. Used in many industries, most notably in the manufacture of pulp, paper, textiles, drinking water, detergents and as a pharmaceutical excipient. Technical grade Sodum Hydroxide products are suitable for general laboratory use.
Specifications
Specifications
Sodium Hydroxide
1
UN1823
2.5 kg
sodium hydroxide
Technical
1310-73-2
HNaO
HEMHJVSKTPXQMS-UHFFFAOYSA-M
[OH-].[Na+]
40.00
Poly Bottle
Chemical Identifiers
1310-73-2
40.00
sodium hydroxide
HNaO
HEMHJVSKTPXQMS-UHFFFAOYSA-M
[OH-].[Na+]
Product Suggestions
Product Suggestions