missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, Flakes, Technical, Spectrum™ Chemical
S1308, 1310-73-2, NaOH
$240.73 - $3314.50
Chemical Identifiers
| CAS | 1310-73-2 |
|---|---|
| Molecular Formula | HNaO |
| Molecular Weight (g/mol) | 40.00 |
| InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
| IUPAC Name | sodium hydroxide |
| SMILES | [OH-].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18606560
|
Spectrum Chemical Mfg Cor
S1308500GM |
500 g | Poly Bottle |
Each for $240.73
|
|
||||
|
18606561
|
Spectrum Chemical Mfg Cor
S130825KG |
2.5 kg | Poly Bottle |
Each for $556.00
|
|
||||
|
18606562
|
Spectrum Chemical Mfg Cor
S130812KG |
12 kg | Poly Pail |
Each for $1,241.93
|
|
||||
|
18606563
|
Spectrum Chemical Mfg Cor
S130850KGBL |
50 kg | Fiber Drum |
Each for $3,314.50
|
|
||||