missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 1% (w/v) (10 g/L), Ricca Chemical
Supplier: Ricca Chemical Company 724316
Specifications
Sodium Hydroxide Solution | |
97.0,0.97 | |
HNaO | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium;hydroxide | |
14798 | |
Natural Poly Bottle | |
500 mL |
1310-73-2,7732-18-5 | |
100.0,1.01 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
[OH-].[Na+] | |
39.997 | |
CHEBI:32145 | |
Aqueous Solution | |
1% (w/v),10g/L |
Chemical Identifiers
1310-73-2 | |
39.997 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, causticShow More | |
CHEBI:32145 | |
[OH-].[Na+] |
HNaO | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
14798 | |
sodium;hydroxide |