Learn More
StatLabâ„¢ SafeTech Vapor Monitor
Vapor monitoring is important to ensure the safety of employees and comply with OSHA guidelines.
$123.49 - $123.49
Specifications
For Use With (Equipment) Laboratory
Type Single Gas Detector
View More Specs
Products 1
Catalog Number Mfr. No. Components Measured Price Quantity  
Catalog Number Mfr. No. Components Measured Price Quantity  
STLVMBFORM
View Documents
StatLabâ„¢
VMBFORM
Formaldehyde
Each for $123.49
Only null left
Description
Description
  • StatLabâ„¢ makes it easy to monitor vapor levels in your laboratory
  • Simply send the badge in and access your results via our website
  • Includes prepaid analysis for 8-hour or 15-minute exposure readings
  • Complies with OSHA standards for vapor exposure in the workplace
Specifications
Specifications
Laboratory
Single Gas Detector
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'