Learn More
(S)-(-)-1,1'-Bi-2-naphthol, 99%
CAS: 18531-99-2 | C20H14O2 | 286.32 g/mol
Supplier: Thermo Scientific Chemicals 224310050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Environmental benefits include:
- Renewable Energy
Specifications
(S)-(-)-1, 1'-Bi-2-naphthol | |
18531-99-2 | |
100.0 | |
White to Gray | |
98.5% min. (HPLC) | |
C20H14O2 | |
5 g | |
15, 1230 | |
1,1'-bi-2-naphthol, s---1,1'-bi-2-naphthol, r-+-1,1'-bi-2-naphthol, 1,1'-binaphthalene-2,2'-diol, binol, 1,1'-binaphthyl-2,2'-diol, 2,2'-dinaphthol, beta-binaphthol, r-1,1'-bi-2-naphthol, s-1,1'-binaphthalene-2,2'-diol | |
PPTXVXKCQZKFBN-UHFFFAOYSA-N | |
1-(2-hydroxynaphthalen-1-yl)naphthalen-2-ol | |
11762 | |
99% |
99% | |
98.5 | |
205°C to 211°C | |
Authentic | |
Glass bottle | |
HOC10H6C10H6OH | |
16,132 | |
-35.5 | |
Solubility in water: insoluble | |
C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)O)O | |
286.32 | |
286.32 | |
Crystalline Powder or Crystals |
Chemical Identifiers
18531-99-2 | |
1,1'-bi-2-naphthol, s---1,1'-bi-2-naphthol, r-+-1,1'-bi-2-naphthol, 1,1'-binaphthalene-2,2'-diol, binol, 1,1'-binaphthyl-2,2'-diol, 2,2'-dinaphthol, beta-binaphthol, r-1,1'-bi-2-naphthol, s-1,1'-binaphthalene-2,2'-diolShow More | |
1-(2-hydroxynaphthalen-1-yl)naphthalen-2-ol |
PPTXVXKCQZKFBN-UHFFFAOYSA-N | |
11762 | |
C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)O)O |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
RUO – Research Use Only