Learn More
(R)-(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid, 99%
CAS: 20445-31-2 | C10H9F3O3 | 234.17 g/mol
Supplier: Thermo Scientific Chemicals 312222500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chiral derivatization reagentSpecifications
(R)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid | |
20445-31-2 | |
White | |
>110°C | |
Glass bottle | |
C6H5C(OCH3)(CF3)CO2H | |
250 mg | |
15,6365 | |
+-mtpa, unii-27o5l9t1wm, r-moshers acid, +-mosher's acid, 2r-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid, r-+-alpha-methoxy-alpha-trifluoromethylphenylacetic acid, r-mtpa, r-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid, r-+-a-methoxy-a-trifluoromethylphenylacetic acid, r-3,3,3-trifluoro-2-methoxy-2-phenyl-propionic acid | |
COC(C(O)=O)(C1=CC=CC=C1)C(F)(F)F | |
234.17 | |
234.17 | |
Crystalline Low Melting Solid |
99%(e.e.>99.5%) | |
46.0°C to 49.0°C | |
116.0°C to 118.0°C (1.5 mmHg) | |
Authentic | |
C10H9F3O3 | |
MFCD00004184 | |
11,329 | |
+ 69.00 | |
JJYKJUXBWFATTE-UHFFFAOYNA-N | |
(2R)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid | |
2723917 | |
99%(e.e.>99.5%) |
Chemical Identifiers
20445-31-2 | |
234.17 | |
JJYKJUXBWFATTE-UHFFFAOYNA-N | |
2723917 | |
COC(C(O)=O)(C1=CC=CC=C1)C(F)(F)F |
C10H9F3O3 | |
MFCD00004184 | |
+-mtpa, unii-27o5l9t1wm, r-moshers acid, +-mosher's acid, 2r-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid, r-+-alpha-methoxy-alpha-trifluoromethylphenylacetic acid, r-mtpa, r-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid, r-+-a-methoxy-a-trifluoromethylphenylacetic acid, r-3,3,3-trifluoro-2-methoxy-2-phenyl-propionic acid | |
(2R)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 243-829-5
Recommended Storage : Refrigerator +4°C
RUO – Research Use Only