missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-Isopropyl-2,5-dimethoxy-3,6-dihydropyrazine, 98%

CAS: 109838-85-9 | C9H16N2O2 | 184.239 g/mol

Supplier:  Thermo Scientific Chemicals H56077AA

 View more versions of this product

Catalog No. AAH56077AA

Only null left
Explore available promotions

Description

Description

(R)-3-Isopropyl-2,5-dimethoxy-3,6-dihydropyrazineChiral auxiliary for the synthesis of α-amino acids. It is an inhibitor with antitumor activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-3-Isopropyl-2,5-dimethoxy-3,6-dihydropyrazineChiral auxiliary for the synthesis of α-amino acids. It is an inhibitor with antitumor activity.

Solubility
Soluble in Chloroform and Ethanol

Notes
Store at -20°C. Keep away from oxidizing agents. Hygroscopic. Keep under inert atmosphere.
Greener Choice Claims
  • Environmental benefits include:
    • Recyclable
Specifications
Show More
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only