missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-Isopropyl-2,5-dimethoxy-3,6-dihydropyrazine, 98%

Catalog No. p-7037361
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH56077XF 1 mL
AAH56077AA 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH56077XF Supplier Thermo Scientific Chemicals Supplier No. H56077XF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 109838-85-9 | C9H16N2O2 | 184.239 g/mol

(R)-3-Isopropyl-2,5-dimethoxy-3,6-dihydropyrazineChiral auxiliary for the synthesis of α-amino acids. It is an inhibitor with antitumor activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-3-Isopropyl-2,5-dimethoxy-3,6-dihydropyrazineChiral auxiliary for the synthesis of α-amino acids. It is an inhibitor with antitumor activity.

Solubility
Soluble in Chloroform and Ethanol

Notes
Store at -20°C. Keep away from oxidizing agents. Hygroscopic. Keep under inert atmosphere.

Chemical Identifiers

CAS 109838-85-9
Molecular Formula C9H16N2O2
Molecular Weight (g/mol) 184.239
MDL Number MFCD00040565
InChI Key FCFWEOGTZZPCTO-MRVPVSSYSA-N
Synonym r-2,5-dihydro-3,6-dimethoxy-2-isopropylpyrazine, r-2-isopropyl-3,6-dimethoxy-2,5-dihydropyrazine, pyrazine, 2,5-dihydro-3,6-dimethoxy-2-1-methylethyl-, 2r, r-2,5-dimethoxy-3-isopropyl-3,6-dihydropyrazine, 2r-2,5-dihydro-3,6-dimethoxy-2-isopropylpyrazine, 2r-2-isopropyl-3,6-dimethoxy-2,5-dihydropyrazine, r-2,5-dihydro-3,6-dimethoxy-2-isopropylpiperazine, 2r-3,6-dimethoxy-2-propan-2-yl-2,5-dihydropyrazine, 2r---2,5-dihydro-3,6-dimethoxy-2-isopropylpyrazine
PubChem CID 736066
IUPAC Name (2R)-3,6-dimethoxy-2-propan-2-yl-2,5-dihydropyrazine
SMILES CC(C)C1C(=NCC(=N1)OC)OC

Specifications

Density 1.028
Flash Point 85°C (185°F)
Quantity 1 mL
Beilstein 3542790
Solubility Information Soluble in Chloroform and Ethanol
Optical Rotation −102° (c=1 in Ethanol)
Formula Weight 184.24
Percent Purity 98%
Chemical Name or Material (R)-3-Isopropyl-2,5-dimethoxy-3,6-dihydropyrazine
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only