missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(-)-2,2-Dimethyl-1,3-dioxolane-4-methanol, 98%, Thermo Scientificâ„¢
$214.10 - $214.10
Chemical Identifiers
CAS | 14347-78-5 |
---|---|
Molecular Formula | C6H12O3 |
Molecular Weight (g/mol) | 132.16 |
MDL Number | MFCD00003213 |
InChI Key | RNVYQYLELCKWAN-RXMQYKEDSA-N |
Synonym | r---2,2-dimethyl-1,3-dioxolane-4-methanol, r-2,2-dimethyl-1,3-dioxolan-4-yl methanol, 4r-2,2-dimethyl-1,3-dioxolan-4-yl methanol, r-solketal, r---1,2-o-isopropylideneglycerol, r-2,2-dimethyl-1,3-dioxolane-4-methanol, 2,3-o-isopropylidene-sn-glycerol, d-acetone glycerol, l---1,2-isopropylideneglycerol, r---2,3-o-isopropylideneglycerolShow More |
PubChem CID | 736056 |
IUPAC Name | [(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]methanol |
SMILES | CC1(OCC(O1)CO)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC221330010
|
Thermo Scientific Chemicals
221330010 |
1g | Glass bottle |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | ||||||||||
AC221330050
|
Thermo Scientific Chemicals
221330050 |
5g | Glass bottle |
Each for $214.10
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
14347-78-5 | |
132.16 | |
RNVYQYLELCKWAN-RXMQYKEDSA-N | |
736056 | |
CC1(OCC(O1)CO)C |
C6H12O3 | |
MFCD00003213 | |
r---2,2-dimethyl-1,3-dioxolane-4-methanol, r-2,2-dimethyl-1,3-dioxolan-4-yl methanol, 4r-2,2-dimethyl-1,3-dioxolan-4-yl methanol, r-solketal, r---1,2-o-isopropylideneglycerol, r-2,2-dimethyl-1,3-dioxolane-4-methanol, 2,3-o-isopropylidene-sn-glycerol, d-acetone glycerol, l---1,2-isopropylideneglycerol, r---2,3-o-isopropylideneglycerolShow More | |
[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]methanol |
Specifications
14347-78-5 | |
100.0 | |
72.0°C to 73.0°C (8.0mmHg) | |
Authentic | |
Glass bottle | |
1.4330 to 1.4350 | |
1g | |
1.06 | |
- 14.00 (20.00°C neat) | |
r---2,2-dimethyl-1,3-dioxolane-4-methanol, r-2,2-dimethyl-1,3-dioxolan-4-yl methanol, 4r-2,2-dimethyl-1,3-dioxolan-4-yl methanol, r-solketal, r---1,2-o-isopropylideneglycerol, r-2,2-dimethyl-1,3-dioxolane-4-methanol, 2,3-o-isopropylidene-sn-glycerol, d-acetone glycerol, l---1,2-isopropylideneglycerol, r---2,3-o-isopropylideneglycerol | |
RNVYQYLELCKWAN-RXMQYKEDSA-N | |
[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]methanol | |
736056 | |
98% |
97.5 | |
1.0600g/mL | |
80°C | |
97.5% min. (GC) | |
C6H12O3 | |
MFCD00003213 | |
19, III, 646 | |
10, 3236 | |
− 14.00 | |
Solubility in water: miscible | |
CC1(OCC(O1)CO)C | |
132.16 | |
132.16 | |
(R)-(-)-2, 2-Dimethyl-1, 3-dioxolane-4-methanol |
RUO – Research Use Only