missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(-)-2,2-Dimethyl-1,3-dioxolane-4-methanol, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 221330010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| (R)-(-)-2, 2-Dimethyl-1, 3-dioxolane-4-methanol | |
| 97.5 | |
| 1.0600g/mL | |
| 80°C | |
| 97.5% min. (GC) | |
| C6H12O3 | |
| MFCD00003213 | |
| 19, III, 646 | |
| 10, 3236 | |
| − 14.00 | |
| Solubility in water: miscible | |
| CC1(OCC(O1)CO)C | |
| 132.16 | |
| 132.16 |
| 14347-78-5 | |
| 100.0 | |
| 72.0°C to 73.0°C (8.0mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.4330 to 1.4350 | |
| 1g | |
| 1.06 | |
| - 14.00 (20.00°C neat) | |
| r---2,2-dimethyl-1,3-dioxolane-4-methanol, r-2,2-dimethyl-1,3-dioxolan-4-yl methanol, 4r-2,2-dimethyl-1,3-dioxolan-4-yl methanol, r-solketal, r---1,2-o-isopropylideneglycerol, r-2,2-dimethyl-1,3-dioxolane-4-methanol, 2,3-o-isopropylidene-sn-glycerol, d-acetone glycerol, l---1,2-isopropylideneglycerol, r---2,3-o-isopropylideneglycerol | |
| RNVYQYLELCKWAN-RXMQYKEDSA-N | |
| [(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]methanol | |
| 736056 | |
| 98% |
RUO â Research Use Only