Learn More
Corning™ PYREX™ VISTA™ Short Neck Round Bottom Boiling Flask, Standard Taper Joint
GREENER_CHOICE
Designed for great shock resistance
Supplier:  Corning™ 70320250
 View more versions of this product
Catalog No. 07-250-082
Only null left
Explore available promotions
Description
Description
These round bottom boiling flasks have full length outer standard taper joints, but with shorter necks. They are manufactured from Code 7740 borosilicate glass. Wall thicknesses are controlled to provide balance between thermal expansion and mechanical strength for greater shock resistance. The round bottom boiling flasks are designed to ASTM E-1403. Adapter Cat. Nos. 8800, 8820, and 8825 are used to connect flasks with different size joints, thus allowing numerous combinations for diverse assemblies.
Compliance
Designed to ASTM E-1403
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications
Specifications
250 mL
Short
2/Pk.
Reusable
Borosilicate Glass
4/Cs.
24/40
Round
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Accessories