Learn More
Corning™ PYREX™ Reducing Glass Joint Adapters
For attaching apparatus with one standard taper joint size to equipment having a different size standard taper joint
Supplier:  Corning™ 88002419
 View more versions of this product
Catalog No. 01-035D
Only null left
Explore available promotions
Description
Description
For attaching apparatus with one standard taper joint size to equipment having a different size standard taper joint
With an outer standard taper joint at the top and a smaller inner standard taper joint at the bottom.
Greener Choice Claims
  • Environmental benefits include:
    • Source Reduction
Specifications
Specifications
For attaching apparatus with one joint size to equipment having a different size joint
105 mm
Reducing Adapter
4.13 in.
Borosilicate Glass
19/38 Inner Bottom, 24/40 Outer Top
Product Suggestions
Product Suggestions