Learn More
Corning™ PYREX™ Reducing Glass Joint Adapters
For attaching apparatus with one standard taper joint size to equipment having a different size standard taper joint
Supplier: Corning™ 88003424
Description
For attaching apparatus with one standard taper joint size to equipment having a different size standard taper joint
With an outer standard taper joint at the top and a smaller inner standard taper joint at the bottom.
Greener Choice Claims
- Environmental benefits include:
- Recyclable
Specifications
For attaching apparatus with one joint size to equipment having a different size joint | |
110 mm | |
Reducing Adapter |
4.33 in. | |
Borosilicate Glass | |
24/40 Inner Bottom, 34/45 Outer Top |