Learn More
Corning™ PYREX™ Graham Condensers with Standard Taper Inner and Outer Joints
GREENER_CHOICE
For use in vacuum distillation
Supplier:  Corning™ 2560300
 View more versions of this product
Catalog No. 07-728A
Only null left
Explore available promotions
Description
Description
For use in vacuum distillation
This condenser has a high surface area per unit length of jacket. Primarily used in vacuum ­distillations. With the outer joint at the top and the inner standard taper joint at the bottom with a drip tip. The tubulation O.D. is approximately 10 mm.
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications
Specifications
11.81 in.
For vacuum distillations
Graham Condenser
41 mm
300 mm
1.61 in.
24/40
450 mm
0.39 in.
17.71 in.
Borosilicate Glass
10 mm
Product Suggestions
Product Suggestions