Learn More
Corning™ PYREX™ Filter Tubes
Used to collect filtrate
Supplier: Corning™ 948032
Description
The constricted lower end is approximately 8 mm O.D. by 75 mm long on all sizes. For use with PYREX brand crucibles with fritted discs listed under Cat. Nos. 32940 and 32960. Also for use with Gooch Âcrucibles or directly with porcelain plates.
Specifications
1.26 in. | |
32 mm | |
Borosilicate Glass | |
Pyrex gooch fritted discs crucibles and porcelain plates | |
16 cm |
0.31 in. | |
0.8 cm | |
Used to collect the filtrate | |
6.29 in. | |
Filter Thimble |