Learn More
Promega ProNex Size-Selective Purification System, 125ml DFS Item
The ProNex Size-Selective Purification System enables the rapid and efficient magnetic resin-based purification of dsDNA for NGS, PCR and general molecular biology applications.
Supplier:  Promega NG2002
Catalog No. PRNG2002
Only null left
Explore available promotions
Description
Description
  • Superior Size Selection and Recovery for NGS and PCR
  • High specificity of size selection with low carryover of unwanted DNA
  • Exceptional DNA recovery
  • Fast magnetic response time and low viscosity for accurate pipetting
Specifications
Specifications
125 mL
Product Suggestions
Product Suggestions