Learn More
Promega SV Total RNA Isolation System
A fast, simple technique for preparation of intact total RNA from tissues, cultured cells and white blood cells in as little as one hour.
Supplier: Promega Z3101
Includes:
- SV Total RNA Isolation System: Spin column assemblies and elution tubes, DNase I (lyophilized), SV RNA lysis buffer, SV RNA dilution buffer, β-mercaptoethanol, MnCl2 (0.09M), Yellow core buffer, SV DNase stop solution, SV RNA wash solution, Nuclease-free water
- SV Total RNA Isolation Start-Up Kit (110V): SV Total RNA Isolation System, Vacuum Adapters, Welch™ Vacuum Pump for North America Electrical, Vac-Man™ Laboratory Vacuum Manifold (20-sample capacity)
Description
- Extracts RNA from Tissues, Cultured Cells and White Blood Cells
- Rapid isolation of high yields of total RNA without the use of hazardous compounds
- Choice of microcentrifugation (spin) or vacuum purification methods
- Purified RNA suitable for all routine molecular biology applications, including RT-PCR
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
15°C to 30°C | |
Molecular biology applications, including RT-PCR and northern blotting | |
10 Preps |
Spin Column | |
Collection Tubes 2 x 5, Nuclease-free Water 1 x 1,250μL, elution tubes 2 x 5, RNA Lysis Buffer (RLA) 1 x 10mL, RNA dilution Buffer (RDA) 1 x 4mL, RNA wash solution (RWA) 1 x 11.8mL, DNase stop solution (DSA) 1 x 5.3mL, yellow core Buffer |