Learn More
Promega Restriction Enzymes - Pvu I (D) DFS Item
Active and capable of digestion directly in GoTaq™ Green Master Mix. (CG AT/CG GC\TA GC)
Supplier:  Promega R6321
 View more versions of this product
Catalog No. PR-R6321
Only null left
Explore available promotions
Description
Description
  • Active and capable of digestion directly in GoTaq(R) Green Master Mix
Specifications
Specifications
2 to 10 U/μL
37°C
Active and capable of digestion directly in GoTaq™ Green Master Mix
-30°C to -10°C
CGAT.CG GC.TAGC
Liquid
1 x 1mL 10X Buffer D, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 100U PvuI
7.3
10mM Tris HCl (pH 7.3 at 25°C), 50mM NaCl, 0.1% Triton™ X-100, 1mM DTT, 0.1mM EDTA, 0.5mg/mL BSA, 50% Glycerol
100 U
PvuI
Product Suggestions
Product Suggestions