Learn More
Praseodymium(III) chloride hydrate, REacton™, 99.9% (REO)
CAS: 19423-77-9 | Cl3Pr | 247.26 g/mol
Supplier: Thermo Scientific Chemicals 01123722
Description
Aqueous solutions of praseodymium(III) chloride can be used to prepare insoluble praseodymium(III) compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Environmental benefits include:
- Recyclable
Specifications
Praseodymium(III) chloride hydrate | |
19423-77-9 | |
100 g | |
Cl3Pr | |
14,7712 | |
LHBNLZDGIPPZLL-UHFFFAOYSA-K | |
praseodymium(3+) trichloride | |
247.27 (Anhydrous) | |
Reacton™ |
115°C | |
Crystalline Aggregates | |
(REO) | |
MFCD00149828 | |
Soluble in water and alcohol | |
[Cl-].[Cl-].[Cl-].[Pr+3] | |
247.26 | |
99.9% | |
Odorless |
Chemical Identifiers
19423-77-9 | |
247.26 | |
LHBNLZDGIPPZLL-UHFFFAOYSA-K | |
[Cl-].[Cl-].[Cl-].[Pr+3] |
Cl3Pr | |
MFCD00149828 | |
praseodymium(3+) trichloride |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
EINECSNumber : 233-794-4
RTECSNumber : TU0175000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only