Learn More
Praseodymium(III) chloride hydrate, REacton™, 99.9% (REO)
CAS: 19423-77-9 | Cl3Pr | 247.26 g/mol
$149.33 - $323.05
Chemical Identifiers
| CAS | 19423-77-9 |
|---|---|
| Molecular Formula | Cl3Pr |
| Molecular Weight (g/mol) | 247.26 |
| MDL Number | MFCD00149828 |
| InChI Key | LHBNLZDGIPPZLL-UHFFFAOYSA-K |
| IUPAC Name | praseodymium(3+) trichloride |
| SMILES | [Cl-].[Cl-].[Cl-].[Pr+3] |
Description
Aqueous solutions of praseodymium(III) chloride can be used to prepare insoluble praseodymium(III) compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 19423-77-9 | |
| 247.26 | |
| LHBNLZDGIPPZLL-UHFFFAOYSA-K | |
| [Cl-].[Cl-].[Cl-].[Pr+3] |
| Cl3Pr | |
| MFCD00149828 | |
| praseodymium(3+) trichloride |
Specifications
| 115°C | |
| Crystalline Aggregates | |
| Cl3Pr | |
| 14,7712 | |
| LHBNLZDGIPPZLL-UHFFFAOYSA-K | |
| praseodymium(3+) trichloride | |
| 247.27 (Anhydrous) | |
| Reacton | |
| Praseodymium(III) chloride hydrate |
| 19423-77-9 | |
| 25 g | |
| MFCD00149828 | |
| Soluble in water and alcohol | |
| [Cl-].[Cl-].[Cl-].[Pr+3] | |
| 247.26 | |
| 99.9% | |
| Odorless | |
| (REO) |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
EINECSNumber : 233-794-4
RTECSNumber : TU0175000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only