missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydrogen Phthalate, 99.95 to 100.05%, ACS, MilliporeSigmaâ„¢
$310.96 - $310.96
Chemical Identifiers
CAS | 877-24-7 |
---|---|
Molecular Formula | C8H5KO4 |
Molecular Weight (g/mol) | 204.222 |
InChI Key | IWZKICVEHNUQTL-UHFFFAOYSA-M |
Synonym | potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasicShow More |
PubChem CID | 23676735 |
IUPAC Name | potassium;2-carboxybenzoate |
SMILES | C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+] |
Chemical Identifiers
877-24-7 | |
204.222 | |
potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasicShow More | |
potassium;2-carboxybenzoate |
C8H5KO4 | |
IWZKICVEHNUQTL-UHFFFAOYSA-M | |
23676735 | |
C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+] |
Specifications
877-24-7 | |
500 g | |
C8H5KO4 | |
IWZKICVEHNUQTL-UHFFFAOYSA-M | |
potassium;2-carboxybenzoate | |
23676735 | |
Potassium Hydrogen Phthalate, Secondary Reference Material For Alkalimetry, Traceable To NIST Standard Reference Material (SRM) Certipureâ„¢ |
Crystals | |
99.95 to 100.05% | |
potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasic | |
C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+] | |
204.222 | |
204.22 |
Safety and Handling
Recommended Storage : Store at +15°C to +25°C