missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydrogen Phthalate, 99.95 to 100.05%, ACS, MilliporeSigma™

Supplier:  Technidata PX14761

 View more versions of this product

Catalog No. MPX14761


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Potassium Hydrogen Phthalate
877-24-7
500 g
C8H5KO4
IWZKICVEHNUQTL-UHFFFAOYSA-M
potassium;2-carboxybenzoate
23676735
Secondary Reference Material For Alkalimetry, Traceable To NIST Standard Reference Material (SRM) Certipure™
Crystals
99.95 to 100.05%
potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasic
C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
204.222
204.22

Chemical Identifiers

877-24-7
204.222
potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasic
potassium;2-carboxybenzoate
C8H5KO4
IWZKICVEHNUQTL-UHFFFAOYSA-M
23676735
C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at +15°C to +25°C

SDS
Documents

Documents

Promotions

Promotions