missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexafluorosilicate, 99.999% (metals basis)

CAS: 16871-90-2 | F6K2Si | 220.27 g/mol

Supplier:  Thermo Scientific Chemicals 04449530

 View more versions of this product

Catalog No. AA4449530


Only null left
Explore available promotions

Description

Description

Potassium hexafluorosilicate is used in the production of porceleins. It is also used for the preservation of timber. It is involved in the manufacturing of ceramics, aluminum and magnesium smelting. In addition, it is used as an intermediate in the manufacturing of optical glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c

H301+H311+H331

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: POTASSIUM FLUOROSILICATE

EINECSNumber : 240-896-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only