missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexafluorosilicate, 99.999% (metals basis)

Catalog No. p-7059342
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4449509 10 g
AA4449518 50 g
AA4449530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4449509 Supplier Thermo Scientific Chemicals Supplier No. 04449509
Only null left

CAS: 16871-90-2 | F6K2Si | 220.27 g/mol

Potassium hexafluorosilicate is used in the production of porceleins. It is also used for the preservation of timber. It is involved in the manufacturing of ceramics, aluminum and magnesium smelting. In addition, it is used as an intermediate in the manufacturing of optical glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16871-90-2
Molecular Formula F6K2Si
Molecular Weight (g/mol) 220.27
MDL Number MFCD00040666
InChI Key UUHPPUWEDCVPCO-UHFFFAOYSA-N
Synonym potassium hexafluorosilicate, potassium fluosilicate, potassium silicofluoride, dipotassium hexafluorosilicate, silicate 2-, hexafluoro-, dipotassium, unii-2610w49xyd, dipotassium hexafluorosilicon 2-, dipotassium hexafluorosilicate 2-, sif6.2k, potassium hexafluorosilicate iv
PubChem CID 61851
IUPAC Name dipotassium;hexafluorosilicon(2-)
SMILES [K+].[K+].F[Si--](F)(F)(F)(F)F

Specifications

Melting Point (decomposition)
Color White
pH 3.4
Physical Form Crystalline
Quantity 10 g
Linear Formula K2SiF6
UN Number UN2655
Merck Index 14,7638
Solubility Information Slightly soluble in water.
Formula Weight 220.28
Percent Purity 99.999%
Sensitivity Moisture sensitive
Density 2.27 g/mL
Assay (metals basis)
Chemical Name or Material Potassium hexafluorosilicate
Show More Show Less
Hazard Category H301+H311+H331
Hazard Statement GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: POTASSIUM FLUOROSILICATE
EINECSNumber 240-896-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only