missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium bromate/Potassium bromide, 0.1N Standardized Solution
KBrO3/KBr
$177.90 - $352.56
Description
Potassium bromate/Potassium bromide is used as an analytical reagent for volumetric analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
KBrO3/KBr | |
MFCD00011359 |
Specifications
Odorless | |
KBrO3/KBr | |
Colorless | |
1 L |
Liquid | |
MFCD00011359 | |
Miscible with water | |
Potassium bromate/Potassium bromide |
Safety and Handling
P201-P202-P281-P308+P313-P501c
H350
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only