missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heathrow Scientific™ Polystyrene Bulk Wrapped Reagent Reservoir

Reagent Reservoirs offer a fresh and modern design with enhanced features to streamline pipetting procedures.
$193.07 - $259.97
Specifications
Color | White |
---|---|
Material | Polystyrene |
Sterility | Non-sterile |
Type | Reagent Reservoir |
Packaging | Bulk Wrapped |
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | ||||||
03449256
![]() |
Heathrow Scientific™
120638 |
10 mL |
Pack of 100 Each for $193.07
|
|
||||||
03449260
![]() |
Heathrow Scientific™
120642 |
25 mL |
Pack of 100 Each for $209.37
|
|
||||||
03449263
![]() |
Heathrow Scientific™
120645 |
50 mL |
Pack of 100 Each for $231.07
|
|
||||||
03449279
![]() |
Heathrow Scientific™
120752 |
55 mL |
Pack of 100 Each for $231.07
|
|
||||||
03449267
![]() |
Heathrow Scientific™
120649 |
100 mL |
Pack of 100 Each for $259.97
|
|
||||||
Description
- Trough design with a v shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
- Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
- Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
- Designed to stay flat on the benchtop and not warp
- Quality certificate available on request
- Disposable
Specifications
White | |
Non-sterile | |
Bulk Wrapped | |
Not Autoclavable | |
Single-use |
Polystyrene | |
Reagent Reservoir | |
100 Pk. | |
1 | |
Convoluted |