Learn More
Plas Labs™ Model 830-CP Compact Glove Box
Plas Lab Model 830-CP Compact Glove Box is designed specifically for use when weighing hazardous materials that must be contained.
Supplier:  Plas Labs™ 830CP21
 View more versions of this product
Catalog No. 11-386-141
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
Offer cleanliness, safety, portability and a draft-free atmosphere for the maximum effectiveness of your balance.
  • Models include access doors; some models also include vents
  • Top section is removable for easy installation of equipment
  • Main chamber holds two purging gas valves
  • Models offer a white leveling tray for easy transferring of liquids, white ambidextrous Hypalon™ gloves for superior chemical and UV resistance, a pressure relief valve that minimizes glove “fight back,” and replaceable HEPA and organic vapor canister filters
  • All clamps are stainless steel and completely adjustable
Order Info
No transfer chamber included
Certifications
Hospital-grade multiple electrical outlet strip is UL listed and CSA certified. CE marked.
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications
Specifications
Glove Box
UL, CSA, CE
29 in.
73.66 cm
34 in.
86 cm
1 Year
35 in.
91.44 cm
HEPA, Organic Vapor
31 in.
78.74 cm
Determining the dry weight or moisture content of aqueous solutions, adhesives, cereals, toxic chemicals, pharmaceuticals, paper, plastics and light radioactive materials
30 in.
17.6 cu. ft.
63 in.
160 cm
Product Suggestions
Product Suggestions