missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenolphthalein, 1% In 60% Isopropanol, pH 8.3 to 10.0 Colorless to Pink, Certified, LabChem™
Supplier: LabChem LC182101
Specifications
Phenolphthalein | |
60,39,1 | |
C20H14O4 | |
Passes Test | |
Certified | |
Carbon dioxide; Carbon monoxide; fume; May release flammable gases | |
8.3 to 10.0 | |
Liquid |
1% In 60% Isopropanol | |
7732-18-5,77-09-8 | |
Soluble in water | |
318.32 | |
Poly Bottle | |
Colorless to Pink | |
500 mL |