Please login to your online account to display your discounted pricing

Phenolphthalein, 1% In 60% Isopropanol, pH 8.3 to 10.0 Colorless to Pink, Certified, LabChem™

$21.14 - $58.54

Chemical Identifiers

Chemical Name or Material Phenolphthalein
CAS 67-63-0
CAS Max % 60
CAS 7732-18-5
CAS Max % 39
CAS 77-09-8
CAS Max % 1
Name Note 1% In 60% Isopropanol, pH 8.3 to 10.0 Colorless to Pink
Purity Grade Certified Grade
Linear Formula C20H14O4
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC182101

 
lab chem
LC182101
500mL Each for $33.74

LC182102

 
lab chem
LC182102
1L Each for $58.54

LC182107

 
lab chem
LC182107
125mL Each for $21.14
Description & Specifications

Specifications

pH 8.3 to 10.0
Purity Grade Certified Grade
Formula Weight 318.32
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of causing cancer.
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.<
Chemical Name or Material Phenolphthalein
Quantity 500mL
CAS Max % 1
EINECS Number 200-661-7
Name Note 1% In 60% Isopropanol
Physical Form Liquid
Linear Formula C20H14O4
CAS 67-63-0
UN Number UN1219
Identification Passes Test
Packaging Poly Bottle
CAS Max % 60
CAS 7732-18-5
Solubility Soluble in water
CAS Max % 39
Recommended Storage Room Temperature
Decomposition Information Carbon dioxide; Carbon monoxide; fume; May release flammable gases
CAS 77-09-8
Color clear, colorless to pink solution
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.<

GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of causing cancer.

Room Temperature