missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenolphthalein, 0.5% In 50% Ethanol, pH 8.3 to 10.0 Colorless to Pink, Certified, LabChem™
Supplier: LabChem LC182002
Description
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
Phenolphthalein | |
49.8,44.8,2.5,2.4,0.5 | |
C20H14O4 | |
Passes Test | |
Certified | |
0.95g/mL | |
0.95g/mL | |
1 L |
0.5% In 50% Ethanol, pH 8.3 to 10.0 Colorless to Pink | |
64-17-5,67-63-0,67-56-1,77-09-8 | |
Soluble in water | |
318.32 | |
Poly Bottle | |
Carbon monoxide; Carbon dioxide | |
Colorless to Pink | |
Liquid |