Please login to your online account to display your discounted pricing

Phenolphthalein, 0.5% In 50% Ethanol, pH 8.3 to 10.0 Colorless to Pink, Certified, LabChem™

$21.00 - $54.26

Chemical Identifiers

Chemical Name or Material Phenolphthalein
CAS 7732-18-5
CAS Max % 49.8
CAS 64-17-5
CAS Max % 44.8
CAS 67-63-0
CAS Max % 2.5
CAS 67-56-1
CAS Max % 2.4
CAS 77-09-8
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC182001

 
lab chem
LC182001
500mL Each for $28.69

LC182002

 
lab chem
LC182002
1L Each for $54.26

LC182007

 
lab chem
LC182007
125mL Each for $21.00
Description & Specifications

Specifications

pH 8.3 to 10.0
Purity Grade Certified Grade
Formula Weight 318.32
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
Flammable liquid and vapor.
May cause cancer.
Suspected of damaging the unborn child (oral).
Causes damage to organs (central nervous system, optic nerve) (oral, dermal).
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, sparks, open flames, hot surfaces.
Keep container tightly closed.
Ground/bond
Chemical Name or Material Phenolphthalein
Quantity 500mL
CAS Max % 2.5
CAS 67-56-1
CAS Max % 2.4
CAS 77-09-8
CAS Max % 0.5
Name Note 0.5% In 50% Ethanol
Physical Form Liquid
Linear Formula C20H14O4
CAS 7732-18-5
UN Number UN1170
Identification Passes Test
Packaging Poly Bottle
CAS Max % 49.8
Viscosity 2.9cSt
CAS 64-17-5
Solubility Soluble in water
Specific Gravity 0.95g/mL
CAS Max % 44.8
Recommended Storage Room Temperature
Decomposition Information Carbon monoxide; Carbon dioxide
Density 0.95g/mL
CAS 67-63-0
Color clear, colorless to pink solution
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, sparks, open flames, hot surfaces.
Keep container tightly closed.
Ground/bond

GHS H Statement
Flammable liquid and vapor.
May cause cancer.
Suspected of damaging the unborn child (oral).
Causes damage to organs (central nervous system, optic nerve) (oral, dermal).

Room Temperature