missing translation for 'onlineSavingsMsg'
Learn More

Phenol:Chloroform:Isoamyl Alcohol (25:24:1), Molecular Biology Grade, Ultrapure

Nucleic acid extraction and purification

$177.55 - $177.55

Specifications

Chemical Name or Material Phenol:Chloroform:Isoamyl Alcohol (25:24:1)
UN Number UN2922
Grade Molecular Biology
Packaging Plastic bottle
Physical Form Liquid
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ75831AE
View Documents
Thermo Fisher Scientific
J75831AE
100 mL
Each for $177.55
Only null left
 
Description

Description

Nucleic acid extraction and purification.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Phenol:Chloroform:Isoamyl Alcohol (25:24:1)
Molecular Biology
Liquid
UN2922
Plastic bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only