Learn More
Palladium, plasma standard solution, Specpure™ Pd 1000μg/mL
Supplier: Thermo Scientific Chemicals 044631AE
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Environmental benefits include:
- Free-of a substance that causes environmental harm
- Non-Toxic
- Ozone-Safe or Safe for the Ozone Layer
- Recyclable
- Recycled Content
- Renewable Energy
- Renewable Materials
Specifications
Palladium, plasma standard solution | |
Liquid | |
MFCD00011167 | |
100 mL |
Pd 1000μg/mL | |
Pd in 5% HNO3 | |
UN3264 | |
Specpure™ |
Chemical Identifiers
MFCD00011167 |
Safety and Handling
GHS H Statement
H314
Causes severe skin burns and eye damage.
P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only