missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Xylene, 99%, for HPLC
CAS: 106-42-3 | C8H10 | 106.17 g/mol
Supplier: Thermo Scientific Chemicals 445680025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Greener Choice Claims
- Environmental benefits include:
- Recyclable
Specifications
p-Xylene | |
106-42-3 | |
0.8660g/mL | |
2.5 L | |
Glass Bottle | |
99% | |
C8H10 | |
MFCD00008556 | |
p-xylene, para-xylene, 1,4-dimethylbenzene, p-methyltoluene, p-dimethylbenzene, p-xylol, benzene, 1,4-dimethyl, 4-xylene, chromar, 4-methyltoluene | |
URLKBWYHVLBVBO-UHFFFAOYSA-N | |
1,4-xylene | |
7809 | |
106.17 | |
HPLC |
13.0°C | |
0.01 max. at 380nm, 0.01 max. at 400nm, 0.05 max. at 350nm, 0.1 max. at 320nm, 1 max. at 294nm | |
138.0°C | |
25°C | |
1.4940 to 1.4960 | |
0.866 | |
C6H4(CH3)2 | |
0.648 mPa.s (20°C) | |
Solubility in water: 0.2g/L (20°C). Other solubilities: soluble in alcohol, ether and most organic, solvents | |
CC1=CC=C(C)C=C1 | |
106.17 | |
CHEBI:27417 | |
99% |
Chemical Identifiers
106-42-3 | |
106.17 | |
URLKBWYHVLBVBO-UHFFFAOYSA-N | |
7809 | |
1,4-xylene |
C8H10 | |
MFCD00008556 | |
p-xylene, para-xylene, 1,4-dimethylbenzene, p-methyltoluene, p-dimethylbenzene, p-xylol, benzene, 1,4-dimethyl, 4-xylene, chromar, 4-methyltolueneShow More | |
CHEBI:27417 | |
CC1=CC=C(C)C=C1 |
Safety and Handling
Danger
RUO – Research Use Only