missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Sexiphenyl, 95%
CAS: 4499-83-6 | C36H26 | 458.604 g/mol
$325.71 - $923.47
Chemical Identifiers
CAS | 4499-83-6 |
---|---|
Molecular Formula | C36H26 |
Molecular Weight (g/mol) | 458.604 |
MDL Number | MFCD00039559 |
InChI Key | ZEMDSNVUUOCIED-UHFFFAOYSA-N |
Synonym | p-sexiphenyl, p-hexaphenyl, p-quaterphenyl, 4,4'-diphenyl, 1-phenyl-4-4-4-4-phenylphenyl phenyl phenyl benzene, p-sexiphenylene, acmc-209k0d, phthalocyanine lead ii purified by sublimation, 1,1':4',1:4,1'-quaterphenyl, 4,4'-diphenyl, 4,4'-bis 1,1'-biphenyl-4-yl-1,1'-biphenyl |
PubChem CID | 78254 |
IUPAC Name | 1-phenyl-4-[4-[4-(4-phenylphenyl)phenyl]phenyl]benzene |
SMILES | C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=C(C=C3)C4=CC=C(C=C4)C5=CC=C(C=C5)C6=CC=CC=C6 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAH26586MD
|
Thermo Scientific Chemicals
H26586MD |
250 mg |
Each for $325.71
|
|
||||||
AAH2658603
|
Thermo Scientific Chemicals
H2658603 |
1 g |
Each for $923.47
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4499-83-6 | |
458.604 | |
ZEMDSNVUUOCIED-UHFFFAOYSA-N | |
78254 | |
C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=C(C=C3)C4=CC=C(C=C4)C5=CC=C(C=C5)C6=CC=CC=C6 |
C36H26 | |
MFCD00039559 | |
p-sexiphenyl, p-hexaphenyl, p-quaterphenyl, 4,4'-diphenyl, 1-phenyl-4-4-4-4-phenylphenyl phenyl phenyl benzene, p-sexiphenylene, acmc-209k0d, phthalocyanine lead ii purified by sublimation, 1,1':4',1:4,1'-quaterphenyl, 4,4'-diphenyl, 4,4'-bis 1,1'-biphenyl-4-yl-1,1'-biphenyl | |
1-phenyl-4-[4-[4-(4-phenylphenyl)phenyl]phenyl]benzene |
Specifications
4499-83-6 | |
C36H26 | |
250 mg | |
ZEMDSNVUUOCIED-UHFFFAOYSA-N | |
1-phenyl-4-[4-[4-(4-phenylphenyl)phenyl]phenyl]benzene | |
78254 | |
95% |
>400°C | |
MFCD00039559 | |
p-sexiphenyl, p-hexaphenyl, p-quaterphenyl, 4,4'-diphenyl, 1-phenyl-4-4-4-4-phenylphenyl phenyl phenyl benzene, p-sexiphenylene, acmc-209k0d, phthalocyanine lead ii purified by sublimation, 1,1':4',1:4,1'-quaterphenyl, 4,4'-diphenyl, 4,4'-bis 1,1'-biphenyl-4-yl-1,1'-biphenyl | |
C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=C(C=C3)C4=CC=C(C=C4)C5=CC=C(C=C5)C6=CC=CC=C6 | |
458.604 | |
458.59 | |
p-Sexiphenyl |
Safety and Handling
EINECSNumber : 224-799-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only