missing translation for 'onlineSavingsMsg'
Learn More

p-Anisidine hydrochloride, 99%

CAS: 20265-97-8 | C7H9NO·HCl | 159.61 g/mol

Supplier:  Thermo Scientific Chemicals 401301000

 View more versions of this product

Catalog No. AC401301000


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications
Show More
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause an allergic skin reaction.<br/

GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Wash with plenty of soap a

GHS Signal Word: Danger

EINECSNumber : 243-657-0

RTECSNumber : BZ6600000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only