Learn More
Oleamide
CAS: 301-02-0 | C18H35NO | 281.48 g/mol
Supplier: Thermo Scientific Chemicals 467650250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Oleamide | |
66.0°C to 76.0°C | |
25 g | |
FATBGEAMYMYZAF-MDZDMXLPSA-N | |
(9E)-octadec-9-enamide | |
281.48 |
301-02-0 | |
C18H35NO | |
Solubility aq. soln.: insoluble. Other solubilities: soluble in ether, ethanol | |
CCCCCCCC\C=C\CCCCCCCC(N)=O | |
281.48 |
Chemical Identifiers
301-02-0 | |
281.48 | |
(9E)-octadec-9-enamide |
C18H35NO | |
FATBGEAMYMYZAF-MDZDMXLPSA-N | |
CCCCCCCC\C=C\CCCCCCCC(N)=O |
Safety and Handling
GHS H Statement Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Warning
EINECSNumber : 206-103-9
RUO – Research Use Only